Drawers, Transparent
1,9 l, Transparent, 168×235×72 mm
Catalog # 216-4346
Supplier: MULTIROIR
Jump to:
Product Details & Documents
System of modular drawer cabinets made from PS. For well organised storage of many items, on shelves or at a workstation.
- Total visibility in the drawer
- Each drawer can be assembled horizontally and vertically
- Each drawer can divided with separators
Documents
VWR® For Your Space VWR® Lab bric-a-brac
Product Literature
Specifications
- Capacity:1,9 l
- Colour:Transparent
- Seq:01
- W×D×H:168×235×72 mm
Accessories
Product Information
- Catalog #AvailabilityPrice
- Transverse separator for drawer 216-4326 (10002)
Product Information
- Catalog #AvailabilityPrice
- Longitudinal separator for drawer 216-4346 (1901)Longitudinal separator for drawer 216-4346 (1901)216-4327Log in for availability1 * 1 ST - 2,30 €
Product Information
- Catalog #AvailabilityPrice
- Transverse separator for drawer 216-4346 (1902)
Product Information
- Catalog #AvailabilityPrice
- Semi-transverse separator for drawer 216-4326 (10003)Semi-transverse separator for drawer 216-4326 (10003)216-4344Log in for availability1 * 1 ST - 2,50 €
Product Information
- Catalog #AvailabilityPrice
- Longitudinal separator for drawer 216-4349 (7001)Longitudinal separator for drawer 216-4349 (7001)216-4339Log in for availability1 * 1 ST - 6,60 €
Product Information
- Catalog #AvailabilityPrice
- Transverse separator for drawer 216-4349 (7002)
Product Information
- Catalog #AvailabilityPrice
- Semi-transverse separator for drawer 216-4346 (1903)Semi-transverse separator for drawer 216-4346 (1903)216-4329Log in for availability1 * 1 ST - 0,90 €
Product Information
- Catalog #AvailabilityPrice
- Semi-transverse separator for drawer 216-4347 (2003)Semi-transverse separator for drawer 216-4347 (2003)216-4338Log in for availability1 * 1 ST - 0,80 €
Product Information
- Catalog #AvailabilityPrice
- Transverse separator for drawer 216-4347 (2002)
Product Information
- Catalog #AvailabilityPrice
- Longitudinal separator for drawer 216-4347 (2001)Longitudinal separator for drawer 216-4347 (2001)216-4336Log in for availability1 * 1 ST - 2,30 €
Product Information
- Catalog #AvailabilityPrice
- Longitudinal separator for drawer 216-4326 (10001)Longitudinal separator for drawer 216-4326 (10001)216-4342Log in for availability1 * 1 ST - 7,30 €
Product Information
- Catalog #AvailabilityPrice
- Semi-transverse separator for drawer 216-4349 (7003)Semi-transverse separator for drawer 216-4349 (7003)216-4341Log in for availability1 * 1 ST - 2,50 €