Orders LinkContactUsLinkComponent
Instrument boxes
Instrument boxes
Silver, 90×180×30 mm
Catalog # AGEN8030
Supplier:   Agencinox
Instrument boxes
Catalog # AGEN8030
Supplier:  Agencinox
Instrument boxes
Instrument boxes
Silver, 90×180×30 mm
Price
35,10 €
Log in for availability
1 * 1 ST
Product Details & Documents
Aluminium, anodised.
  • Smooth surfaces for fast, easy cleaning
  • Coloured
  • Good chemical resistance, very good thermal resistance
Specifications
  • Colour:
    Silver
  • Seq:
    02
  • W×D×H:
    90×180×30 mm