Orders LinkContactUsLinkComponent
Dissecting trays
Dissecting trays
310×200×45 mm
Catalog # NICK625
Supplier:   NICKEL ELECTRO
Dissecting trays
Catalog # NICK625
Supplier:  NICKEL ELECTRO
Dissecting trays
Dissecting trays
310×200×45 mm
Price
57,20 €
Log in for availability
1 * 1 ST
Product Details & Documents
Dissecting trays made of aluminium.
  • Good chemical resistance
  • Good temperature resistance
  • Easy to clean
Specifications
  • Seq:
    01
  • W×D×H:
    310×200×45 mm