Order Entry
Germany
Orders LinkContactUsLinkComponent
 

Clamps

Stainless steel 18/10, 60 mm, 150 mm
Catalog # BOCH5408
Supplier:   BOCHEM
Clamps
Catalog # BOCH5408
Supplier:  BOCHEM
Clamps
Clamps

Stainless steel 18/10, 60 mm, 150 mm

Price
27,40 €
Log in for availability
1 * 1 ST
Product Details & Documents

Retort clamps made of stainless steel.

  • Jaws are covered with a layer of cork to protect clamped items
  • Angled jaws

 : DIN 12894

Specifications
  • Spanning width:
    60 mm
  • Length:
    150 mm
  • Material:
    Stainless steel 18/10
  • Seq:
    03