Order Entry
Germany
Orders LinkContactUsLinkComponent
Weighing scoops

Weighing scoops

Weighing scoop, conical shape, 330 mm, 100 mm
Catalog # BOCH3606
Supplier:   BOCHEM
Weighing scoops
Catalog # BOCH3606
Supplier:  BOCHEM
Weighing scoops
Weighing scoops

Weighing scoop, conical shape, 330 mm, 100 mm

Price
45,00 €
Log in for availability
1 * 1 ST
Product Details & Documents

These conical weighing scoops are made of 18/10 stainless steel.

Specifications
  • Width:
    100 mm
  • Description:
    Weighing scoop, conical shape
  • Length:
    330 mm
  • Seq:
    03
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.