Order Entry
Germany
Orders LinkContactUsLinkComponent
Tissue capsules

Tissue capsules

White, 38×10 mm
Catalog # 720-0206
Supplier:   Simport Scientific
Tissue capsules
Catalog # 720-0206
Supplier:  Simport Scientific
Tissue capsules
Tissue capsules

White, 38×10 mm

Price
481,00 €
Log in for availability
1 * 1.000 ST
Product Details & Documents

These capsules made of white PP can hold tissue during processing.

  • Frosted area for writing sample ID
  • Lid snaps securely on to the base
Specifications
  • Type:
    38×10 mm
  • Colour:
    White