Orders LinkContactUsLinkComponent
Turbidity standards, Stablcal®
Turbidity standards, Stablcal®
Stablcal® stabilised formazine standard, 1,0 NTU, 1 L
Catalog # 708-0079
Supplier:   Hach
Turbidity standards, Stablcal®
Catalog # 708-0079
Supplier:  Hach
CAS Number:  
Turbidity standards, Stablcal®
Turbidity standards, Stablcal®
Stablcal® stabilised formazine standard, 1,0 NTU, 1 L
CAS Number:  
Price
251,00 €
Log in for availability
1 * 1 L
Product Details & Documents
Stablcal® turbidity standards for all turbidimeters. USEPA accepted for calibration purposes. Stablcal® stabilised formazine turbidity standards are true formazine dilutions developed for use in any turbidimeter.
  • Precise, pre-mixed formazine solutions
  • Based on formazine primary standards
  • Enhanced stability saves time and labour
  • Confidence in results
Documents
Specifications
Specifications
  • Pk:
    1 L
  • Turbidity:
    1,0 NTU
  • Description:
    Stablcal® stabilised formazine standard