Order Entry
Germany
Orders LinkContactUsLinkComponent
Tissue processing cassettes, Histosette I

Tissue processing cassettes, Histosette I

Pink
Catalog # DELT454140
Supplier:   DELTALAB
Tissue processing cassettes, Histosette I
Catalog # DELT454140
Supplier:  DELTALAB
Tissue processing cassettes, Histosette I

Pink

Price
164,00 €
Log in for availability
1 * 500 ST
Product Details & Documents

Made of acetal polymer. With one-piece integral lid. To lock the cassette, just snap apart the lid, turn it round and lock it into the base of the cassette.

  • Writing area at 30° angle

Documents

Specifications
  • Colour:
    Pink
  • Seq:
    01
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.