Order Entry
Germany
Orders LinkContactUsLinkComponent
Ammonia solution ∼32%, GPR RECTAPUR®

Ammonia solution ∼32%, GPR RECTAPUR®

Glass bottle, Safebreak®, 2,5 L
Artikelnummer 21192.323
Corrosive
Irritant
Environment
Hersteller:   VWR Chemicals
Ammonia solution ∼32%, GPR RECTAPUR®
Artikelnummer 21192.323
Hersteller:  VWR Chemicals
CAS-Nummer:  1336-21-6
Ammonia solution ∼32%, GPR RECTAPUR®
Ammonia solution ∼32%, GPR RECTAPUR®

Glass bottle, Safebreak®, 2,5 L

CAS-Nummer:  1336-21-6
Preis
66,10 €
Log in for availability
1 * 2,5 L
Product Details & Documents
Synonyms:  Ammonia aqueous, Ammonium hydroxide solution, Ammonium hydroxide, Ammonia water
  • CAS Number:1336-21-6
  • Formula:NH₃
  • MDL Number:MFCD00066650
  • Molecular Weight:35.05 g/mol
  • Boiling Pt:32…37.7 °C (1013 hPa)
  • Melting Pt:–60…–57.5 °C
  • Density:0.88 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:215-647-6
  • UN:2672
  • ADR:8,III
Specifications

Specification Test Results

  • Assay:
    28 - 33 %
  • Density (20/4):
    0.880 - 0.910
  • Evaporation residue:
    Max. 100 ppm
  • Heavy metals (as Pb):
    Max. 10 ppm
  • Cl (Chloride):
    Max. 10 ppm
  • CO3 (Carbonate):
    Max. 100 ppm
  • SO4 (Sulphate):
    Max. 20 ppm
  • Fe (Iron):
    Max. 1 ppm
  • Pb (Lead):
    Max. 1 ppm

Specifications

  • Pack type:
    Glass bottle, Safebreak®
  • Pk:
    2,5 L

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.