Orders LinkContactUsLinkComponent
Turbidity standards, Stablcal®
Stablcal® stabilised formazine standard, 800 NTU, 500 mL
Catalog # 708-0069
Supplier:   Hach
Turbidity standards, Stablcal®
Catalog # 708-0069
Supplier:  Hach
CAS Number:  
Turbidity standards, Stablcal®
Turbidity standards, Stablcal®
Stablcal® stabilised formazine standard, 800 NTU, 500 mL
CAS Number:  
Price
203,00 €
Log in for availability
1 * 500 mL
Product Details & Documents
Stablcal® turbidity standards for all turbidimeters. USEPA accepted for calibration purposes. Stablcal® stabilised formazine turbidity standards are true formazine dilutions developed for use in any turbidimeter.
  • Precise, pre-mixed formazine solutions
  • Based on formazine primary standards
  • Enhanced stability saves time and labour
  • Confidence in results
Documents
Specifications
Specifications
  • Pk:
    500 mL
  • Turbidity:
    800 NTU
  • Description:
    Stablcal® stabilised formazine standard