Orders LinkContactUsLinkComponent
Turbidity standards, Stablcal®
Stablcal® stabilised formazine standard, <0,1 NTU, 500 mL
  708-0083
 :   Hach
Turbidity standards, Stablcal®
  708-0083
 :  Hach
 :  
Turbidity standards, Stablcal®
Turbidity standards, Stablcal®
Stablcal® stabilised formazine standard, <0,1 NTU, 500 mL
 :  
102,00 €
1 * 500 mL
Stablcal® turbidity standards for all turbidimeters. USEPA accepted for calibration purposes. Stablcal® stabilised formazine turbidity standards are true formazine dilutions developed for use in any turbidimeter.
  • Precise, pre-mixed formazine solutions
  • Based on formazine primary standards
  • Enhanced stability saves time and labour
  • Confidence in results
  • Pk:
    500 mL
  • Turbidity:
    <0,1 NTU
  • Description:
    Stablcal® stabilised formazine standard