Orders LinkContactUsLinkComponent
Nitrogen-nitrate reagent sets, LR
Nitrogen-nitrate reagent sets, LR
Nitrogen nitrate reagent set, 0 - 0,50 mg/l NO₃- N, Cadmium reduction, DR3000, DR2000, 25 ml, PP, 100, 100 Tests
Catalog # HACH2245100
Supplier:   Hach
Nitrogen-nitrate reagent sets, LR
Catalog # HACH2245100
Supplier:  Hach
CAS Number:  
Nitrogen-nitrate reagent sets, LR
Nitrogen-nitrate reagent sets, LR
Nitrogen nitrate reagent set, 0 - 0,50 mg/l NO₃- N, Cadmium reduction, DR3000, DR2000, 25 ml, PP, 100, 100 Tests
CAS Number:  
Price
180,00 €
Log in for availability
1 * 100 Tests
Product Details & Documents
For determination of nitrogen-nitrate by the cadmium reduction method.
Specifications
Specifications
  • Chemical method:
    Cadmium reduction
  • Pk:
    100 Tests
  • Description:
    Nitrogen nitrate reagent set
  • Sample size:
    25 ml
  • Type:
    PP
  • For unit:
    DR3000, DR2000
  • Tests per kit:
    100
  • Range:
    0 - 0,50 mg/l NO₃- N
Specifications