Order Entry
Germany
Orders LinkContactUsLinkComponent
Hydrochloric acid 30%, TECHNICAL

Hydrochloric acid 30%, TECHNICAL

Plastic bottle, 5 L
Catalog # 20251.365
Corrosive
Irritant
Supplier:   VWR Chemicals
Hydrochloric acid 30%, TECHNICAL
Catalog # 20251.365
Supplier:  VWR Chemicals
CAS Number:  7647-01-0
Hydrochloric acid 30%, TECHNICAL
Hydrochloric acid 30%, TECHNICAL

Plastic bottle, 5 L

CAS Number:  7647-01-0
Price
51,70 €
Log in for availability
1 * 5 L
Product Details & Documents
Synonyms:  Hydrochloric acid solution, Muriatic acid
  • CAS Number:7647-01-0
  • Formula:HCl
  • MDL Number:MFCD00011324
  • Molecular Weight:36.46 g/mol
  • Boiling Pt:110 °C (1013 hPa)
  • Melting Pt:–30 °C
  • Density:1.18 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:231-595-7
  • UN:1789
  • ADR:8,II
  • REACH:01-2119484862-27
Specifications

Specification Test Results

  • Assay:
    29 - 31 %

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    5 L

Specifications