Order Entry
Germany
Orders LinkContactUsLinkComponent
EDTA solutions

EDTA solutions

EDTA standard solution, 0,035 N, 100 ml, for hardness determination by drop-count titration, 100 mL
Catalog # HACH2349932
Supplier:   Hach
EDTA solutions
Catalog # HACH2349932
Supplier:  Hach
CAS Number:  
EDTA solutions
EDTA solutions

EDTA standard solution, 0,035 N, 100 ml, for hardness determination by drop-count titration, 100 mL

CAS Number:  
Price
26,10 €
Log in for availability
1 * 100 mL
Product Details & Documents

Documents

Specifications

Specifications

  • Pk:
    100 mL
  • Description:
    EDTA standard solution, 0,035 N, 100 ml, for hardness determination by drop-count titration

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.