Orders LinkContactUsLinkComponent
EDTA solutions
EDTA solutions
EDTA solution, 1 M, 50 ml, for determination of calcium and magnesium hardness in water by the calmagite colorimetric method, Marked dropping bottle, 50 mL
Catalog # HACH2241926
Supplier:   Hach
EDTA solutions
Catalog # HACH2241926
Supplier:  Hach
CAS Number:  
EDTA solutions
EDTA solutions
EDTA solution, 1 M, 50 ml, for determination of calcium and magnesium hardness in water by the calmagite colorimetric method, Marked dropping bottle, 50 mL
CAS Number:  
Price
21,10 €
Log in for availability
1 * 50 mL
Product Details & Documents
Documents
Specifications
Specifications
  • Pack type:
    Marked dropping bottle
  • Pk:
    50 mL
  • Description:
    EDTA solution, 1 M, 50 ml, for determination of calcium and magnesium hardness in water by the calmagite colorimetric method
Specifications