Order Entry
Germany
Orders LinkContactUsLinkComponent
Calcium standard solution

Calcium standard solution

Calcium, Ca, 10000 mg/L, 2% HCl, 16
Catalog # HACH218710
Supplier:   Hach
Calcium standard solution
Catalog # HACH218710
Supplier:  Hach
CAS Number:  
Calcium standard solution
Calcium standard solution

Calcium, Ca, 10000 mg/L, 2% HCl, 16

CAS Number:  
Price
63,30 €
Log in for availability
1 * 16 ST
Product Details & Documents

Standard solution for accuracy checks in calcium hardness determinations.

Documents

Specifications

Specifications

  • Concentration:
    10000 mg/L
  • Matrix:
    2% HCl
  • Pk:
    16
  • Element:
    Calcium
  • Symbol:
    Ca

Specifications