Order Entry
Germany
Orders LinkContactUsLinkComponent
Solid phase extraction cartridges, Bond Elut™ LMS

Solid phase extraction cartridges, Bond Elut™ LMS

Straight barrel cartridge, LMSA, 300 Å, 75 µm, 3 ml, 500 mg
Catalog # 551-0342
Supplier:   VARIAN
Solid phase extraction cartridges, Bond Elut™ LMS
Catalog # 551-0342
Supplier:  VARIAN
Solid phase extraction cartridges, Bond Elut™ LMS

Straight barrel cartridge, LMSA, 300 Å, 75 µm, 3 ml, 500 mg

Price
514,00 €
Log in for availability
1 * 50 ST
Product Details & Documents

Bond Elut™ LMS polymeric sorbent lets you elute without having to add amine modifiers, buffers, or acids. The elimination of secondary interactions means that elution of analytes can be achieved with pure organic solvents or HPLC mobile phase-compatible solvent mixtures of low ionic strength. These characteristics are crucial to allow compatibility with LC/MS or other delicate analytical techniques.

  • Ultra clean styrene-divinylbenzene polymer
  • Optimised 75 μm particle size for reproducible flow
  • High capacity and surface area for efficient extraction
Specifications
  • Particle Size:
    75 µm
  • Pore size:
    300 Å
  • Capacity:
    3 ml
  • Description:
    Straight barrel cartridge
  • Seq:
    5
  • Bed weight:
    500 mg
  • Phase:
    LMSA