Solid phase extraction cartridges, Bond Elut™ LMS
Straight barrel cartridge, LMSA, 300 Å, 75 µm, 3 ml, 100 mg
Catalog # 551-0340
Supplier: VARIAN
Jump to:
Product Details & Documents
Bond Elut™ LMS polymeric sorbent lets you elute without having to add amine modifiers, buffers, or acids. The elimination of secondary interactions means that elution of analytes can be achieved with pure organic solvents or HPLC mobile phase-compatible solvent mixtures of low ionic strength. These characteristics are crucial to allow compatibility with LC/MS or other delicate analytical techniques.
- Ultra clean styrene-divinylbenzene polymer
- Optimised 75 μm particle size for reproducible flow
- High capacity and surface area for efficient extraction
Specifications
- Particle Size:75 µm
- Pore size:300 Å
- Capacity:3 ml
- Description:Straight barrel cartridge
- Bed weight:100 mg
- Phase:LMSA
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



