Order Entry
Germany
Orders LinkContactUsLinkComponent
 

VWR® AVS TITRINORM Buffer Solutions, 20 °C

7,00, ±0,02, Phosphate, Colourless, ISO 17034, Plastic bottle, 500 mL
Catalog # 32096.267
Supplier:   VWR Chemicals
VWR® AVS TITRINORM Buffer Solutions, 20 °C
Catalog # 32096.267
Supplier:  VWR Chemicals
CAS Number:  
VWR® AVS TITRINORM Buffer Solutions, 20 °C
VWR® AVS TITRINORM Buffer Solutions, 20 °C

7,00, ±0,02, Phosphate, Colourless, ISO 17034, Plastic bottle, 500 mL

CAS Number:  
Price
29,20 €
Log in for availability
1 * 500 mL
Product Details & Documents

Solutions prepared from AnalaR® NORMAPUR® grade analytical reagents.

  • Reference temperature 20 °C

  • True analytical value on the label to an accuracy of ±0,02 pH units (pH 1,00 to 10,00); ±0,05 for pH 11,00 and pH 12,00 buffer
  • Traceable to SRM from NIST
  • Batch number and packing date for traceability
  • Certificate of Analysis available at website

Manufactured from analytical grade products presenting high accuracy: 0,02 pH unit from pH 1,00 to 10,00 and 0,05 pH unit for pH 11 and pH 12,0.

 : Certifications: Products with accreditation to ISO 17034 in the table below are produced and certified in accordance with ISO 17034 accredited by BELAC under the certification number 542-RM. All information regarding the use of these CRMs can be found on the Certificate of Analysis.

 : If significantly large volumes are required please contact your local Avantor sales office for a customised offer.

 : Available in different pack sizes: 100 ml, 500 ml, 1 L, 5 L plastic bottles and bag-in-box (5 and 10 L).

Specifications

Specifications

  • Pack type:
    Plastic bottle
  • pH buffer type:
    Phosphate
  • Pk:
    500 mL
  • Colour:
    Colourless
  • pH accuracy:
    ±0,02
  • pH value:
    7,00
  • Accreditation:
    ISO 17034

Specifications