Order Entry
Germany
Orders LinkContactUsLinkComponent
VWR® AVS TITRINORM Buffer Solutions, 20 °C

VWR® AVS TITRINORM Buffer Solutions, 20 °C

7,00, ±0,02, Phosphate, Green, 20 °C, ISO 17034, Bag-in-box, 5 L
Catalog # 32045.375
Supplier:   VWR Chemicals
VWR® AVS TITRINORM Buffer Solutions, 20 °C
Catalog # 32045.375
Supplier:  VWR Chemicals
CAS Number:  
VWR® AVS TITRINORM Buffer Solutions, 20 °C
VWR® AVS TITRINORM Buffer Solutions, 20 °C

7,00, ±0,02, Phosphate, Green, 20 °C, ISO 17034, Bag-in-box, 5 L

CAS Number:  
Price
205,00 €
Log in for availability
1 * 5 L
Product Details & Documents

Solutions prepared from AnalaR® NORMAPUR® grade analytical reagents.

  • Reference temperature 20 °C

  • True analytical value on the label to an accuracy of ±0,02 pH units (pH 1,00 to 10,00); ±0,05 for pH 11,00 and pH 12,00 buffer
  • Traceable to SRM from NIST
  • Batch number and packing date for traceability
  • Certificate of Analysis available at website

Manufactured from analytical grade products presenting high accuracy: 0,02 pH unit from pH 1,00 to 10,00 and 0,05 pH unit for pH 11 and pH 12,0.

Certifications: Certifications: Products with accreditation to ISO 17034 in the table below are produced and certified in accordance with ISO 17034 accredited by BELAC under the certification number 542-RM. All information regarding the use of these CRMs can be found on the Certificate of Analysis.

Ordering information: If significantly large volumes are required please contact your local Avantor sales office for a customised offer.

Packaging: Available in different pack sizes: 100 ml, 500 ml, 1 L, 5 L plastic bottles and bag-in-box (5 and 10 L).

Specifications

Specifications

  • Pack type:
    Bag-in-box
  • pH buffer type:
    Phosphate
  • Pk:
    5 L
  • Reference temperature:
    20 °C
  • Colour:
    Green
  • pH accuracy:
    ±0,02
  • pH value:
    7,00
  • Accreditation:
    ISO 17034

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.