Order Entry
Germany
Orders LinkContactUsLinkComponent
Phenol red solution pH-indicator (pH 6,5 - 8,5)

Phenol red solution pH-indicator (pH 6,5 - 8,5)

Dropping bottle, marked, 100 mL
Catalog # HACH21132
Supplier:   Hach
Phenol red solution pH-indicator (pH 6,5 - 8,5)
Catalog # HACH21132
Supplier:  Hach
CAS Number:  143-74-8
Phenol red solution pH-indicator (pH 6,5 - 8,5)
Phenol red solution pH-indicator (pH 6,5 - 8,5)

Dropping bottle, marked, 100 mL

CAS Number:  143-74-8
Price
25,30 €
Log in for availability
1 * 100 mL
Product Details & Documents

Phenol Red Indicator Solution provides an easy-to-use colorimetric method for determining pH levels for neutral and slightly alkaline waters in the range of 6.5 to 8.5 units. This method uses a color wheel and is especially useful for testing swimming pools and spas.

In addition to these applications, Phenol Red is also used for monitoring ground and river water quality, making it a versatile tool for various environmental water quality testing needs.

Color range: pH 6.5 - 8.5

Synonyms:  3,3-Bis(4-hydroxyphenyl)-3H-benzo[c][1,2]oxathiole 1,1-dioxide
  • CAS Number:143-74-8
  • Formula:C₁₉H₁₄O₅S
  • MDL Number:MFCD00003552
  • Molecular Weight:354.38 g/mol
Specifications

Specifications

  • Pack type:
    Dropping bottle, marked
  • Pk:
    100 mL

Specifications