Order Entry
Germany
Orders LinkContactUsLinkComponent
Copper standard solution

Copper standard solution

Copper standard solution (NIST), 100 mg/l as Cu, 100 mL
Catalog # HACH12842
Supplier:   Hach
Copper standard solution
Catalog # HACH12842
Supplier:  Hach
CAS Number:  
Copper standard solution

Copper standard solution (NIST), 100 mg/l as Cu, 100 mL

CAS Number:  
Price
32,90 €
Log in for availability
1 * 100 mL
Product Details & Documents

Standard solution for accuracy checks in copper determinations.

Specifications

Specifications

  • Concentration:
    100 mg/l as Cu
  • Pk:
    100 mL
  • Description:
    Copper standard solution (NIST)

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.