Orders LinkContactUsLinkComponent
Comparator kits for water testing
Nitrates (low concentration), 0 - 1 mg/l, 50, 1 KIT
  705-0055
 :   Hach
Comparator kits for water testing
  705-0055
 :  Hach
 :  
Comparator kits for water testing
Comparator kits for water testing
Nitrates (low concentration), 0 - 1 mg/l, 50, 1 KIT
 :  
183,00 €
1 * 1 KIT
Qualitative results using traditional colour comparator disc methods.
  • Portable - ideal for field testing
  • Simple fast method
  • Kits contain all required reagents and apparatus in a rugged carrying case
Please note some of the reagents supplied in the kits are hazardous, and appropriate precautions should be taken for use and disposal. Please contact Avantor for details.
  • Pk:
    1 KIT
  • Description:
    Nitrates (low concentration)
  • Tests per kit:
    50
  • Range:
    0 - 1 mg/l
Accessories
  •  
    Description
    Pk
  • HACH11032H
    Ammonium molybdate reagent for phosphate determinations, 100 ml
    1
    1 * 1 ST - 22,60 €