Order Entry
Germany
Orders LinkContactUsLinkComponent
Sodium hydroxide 30% in aqueous solution, TECHNICAL

Sodium hydroxide 30% in aqueous solution, TECHNICAL

Plastic drum, 200 L
Catalog # 28217.554
Corrosive
Supplier:   VWR Chemicals
Sodium hydroxide 30% in aqueous solution, TECHNICAL
Catalog # 28217.554
Supplier:  VWR Chemicals
CAS Number:  1310-73-2
Sodium hydroxide 30% in aqueous solution, TECHNICAL
Sodium hydroxide 30% in aqueous solution, TECHNICAL

Plastic drum, 200 L

CAS Number:  1310-73-2
Price
Inquire For Price
Log in for availability
1 * 200 L
Product Details & Documents
Synonyms:  Caustic soda
  • CAS Number:1310-73-2
  • Formula:NaOH
  • MDL Number:MFCD00003548
  • Molecular Weight:40 g/mol
  • Boiling Pt:119…135 °C (1013 hPa)
  • Melting Pt:9…10 °C
  • Density:1,53 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:215-185-5
  • UN:1824
  • ADR:8,II
Specifications

Specification Test Results

  • Assay:
    Min. 29 %
  • Heavy metals (as Pb):
    Max. 50 ppm

Specifications

  • Pack type:
    Plastic drum
  • Pk:
    200 L

Specifications