Order Entry
Germany
Orders LinkContactUsLinkComponent
Sodium hydroxide 30% in aqueous solution, TECHNICAL

Sodium hydroxide 30% in aqueous solution, TECHNICAL

Plastic bottle, 1 L
Catalog # 28217.292
Corrosive
Supplier:   VWR Chemicals
Sodium hydroxide 30% in aqueous solution, TECHNICAL
Catalog # 28217.292
Supplier:  VWR Chemicals
CAS Number:  1310-73-2
Sodium hydroxide 30% in aqueous solution, TECHNICAL
Sodium hydroxide 30% in aqueous solution, TECHNICAL

Plastic bottle, 1 L

CAS Number:  1310-73-2
Price
32,40 €
Log in for availability
1 * 1 L
Product Details & Documents
Synonyms:  Caustic soda
  • CAS Number:1310-73-2
  • Formula:NaOH
  • MDL Number:MFCD00003548
  • Molecular Weight:40 g/mol
  • Boiling Pt:119…135 °C (1013 hPa)
  • Melting Pt:9…10 °C
  • Density:1,53 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:215-185-5
  • UN:1824
  • ADR:8,II
Specifications

Specification Test Results

  • Assay:
    Min. 29 %
  • Heavy metals (as Pb):
    Max. 50 ppm

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    1 L

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.