Orders LinkContactUsLinkComponent
Ampoule breakers
Ampoule breakers
Ampoule breaker, Voluette ampoules
Catalog # HACH2196800
Supplier:   Hach
Ampoule breakers
Catalog # HACH2196800
Supplier:  Hach
Ampoule breakers
Ampoule breakers
Ampoule breaker, Voluette ampoules
Price
38,30 €
Log in for availability
1 * 1 KIT
Product Details & Documents
These breakers for ampoules are designed to prevent contamination. Breaker snaps ampoule tip off.
Specifications
  • Description:
    Ampoule breaker
  • Seq:
    03
  • For:
    Voluette ampoules