Order Entry
Germany
Orders LinkContactUsLinkComponent
N,N-Dimethylformamide ≥98%, TECHNICAL

N,N-Dimethylformamide ≥98%, TECHNICAL

Plastic bottle, 1 L
Catalog # 23469.298
Flammable
Health
Irritant
Supplier:   VWR Chemicals
N,N-Dimethylformamide ≥98%, TECHNICAL
Catalog # 23469.298
Supplier:  VWR Chemicals
CAS Number:  68-12-2
N,N-Dimethylformamide ≥98%, TECHNICAL
N,N-Dimethylformamide ≥98%, TECHNICAL

Plastic bottle, 1 L

CAS Number:  68-12-2
Price
35,10 €
Log in for availability
1 * 1 L
Product Details & Documents
Synonyms:  Formic acid dimethylamide, Formdimethylamide, Dimethylformamide, DMF
  • CAS Number:68-12-2
  • Formula:HCON(CH₃)₂
  • MDL Number:MFCD00003284
  • Molecular Weight:73.09 g/mol
  • Merck Index:13,03269
  • Boiling Pt:153 °C (1013 hPa)
  • Melting Pt:–60,5 °C
  • Density:0.949 g/cm³ (20 °C)
  • Flash Pt:57.5 °C (DIN 51755) (closed cup)
  • Storage Temperature:Ambient
  • EINECS:200-679-5
  • UN:2265
  • ADR:3,III
  • REACH:01-2119475605-32
Specifications

Specification Test Results

  • Assay:
    Min. 98 %
  • Refractive index (20°C):
    1.429 - 1.431

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    1 L

Specifications