Orders LinkContactUsLinkComponent
Distillation flask, STANHOPE-SETA
Distillation flask, STANHOPE-SETA
200 ml
Catalog # STAN11962-0
Supplier:   STANHOPE SETA
Distillation flask, STANHOPE-SETA
Catalog # STAN11962-0
Supplier:  STANHOPE SETA
Distillation flask, STANHOPE-SETA
Distillation flask, STANHOPE-SETA
200 ml
Price
694,00 €
Log in for availability
1 * 5 ST
Product Details & Documents
For the distillation of petroleum products.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
Complies with ASTM D850.
Specifications
  • Capacity:
    200 ml
  • Seq:
    01