Order Entry
Germany
Orders LinkContactUsLinkComponent
Sampling cans

Sampling cans

Sampling can, Copper, 1500 ml, 450 mm, 38/83 mm
Catalog # STAN14750-0
Supplier:   STANHOPE SETA
Sampling cans
Catalog # STAN14750-0
Supplier:  STANHOPE SETA
Sampling cans
Sampling cans

Sampling can, Copper, 1500 ml, 450 mm, 38/83 mm

Price
Inquire For Price
Log in for availability
1 * 1 ST
Product Details & Documents

Vessels to take samples from non-pressurised bulk storage without contamination of the laboratory equipment.

  • Stainless steel
Specifications
  • Ø:
    38/83 mm
  • Capacity:
    1500 ml
  • Description:
    Sampling can
  • Length:
    450 mm
  • Material:
    Copper
  • Seq:
    02
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.