Orders LinkContactUsLinkComponent
Safety funnels, with handle
Safety funnels, with handle
250 mm
Catalog # AGEN4250
Supplier:   Agencinox
Safety funnels, with handle
Catalog # AGEN4250
Supplier:  Agencinox
Safety funnels, with handle
Safety funnels, with handle
250 mm
Price
141,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
Stainless steel.
  • Excellent chemical resistance
  • Excellent temperature resistance
  • Easy to clean
Documents
Specifications
  • Funnel O-Ø:
    250 mm
  • Seq:
    03