Spatulas
Spatula, analysis type (A), 230 mm
Supplier: Agencinox
Jump to:
Product Details & Documents
Stainless steel ¹⁸/₁₀, anti-magnetic, electro-polishing. These universal spatulas are suitable for many applications.
- Excellent chemical and thermal resistance
- Autoclavable
Dokumente
Specifications
- Description:Spatula, analysis type (A)
- Length:230 mm