VWR®, Clamps
20 - 75 mm
241-0023
Stainless steel. Easy to use, robust clamps.
- Independently adjustable jaws, jaw height 16 and 25 mm
- With two easy action knurled screws for adjusting each of the movable clamping jaws independently of each other
- Spanning width:20 - 75 mm
- Seq:2



