Dropping bottles, LDPE, WHEATON®
3 ml, LDPE, translucent, 8-425
Catalog # 215-8000
Supplier: DWK Life Sciences
Jump to:
Product Details & Documents
LDPE, translucent, with white PP cap.
- Good chemical resistance, heat resistant up to 80 °C
- Bottle dispenses one drop at a time
- Dropper tip snaps firmly in place and is easily removed for filling
With extended controlled dropper tip and dropper tip cap. These dropping bottles provide reliable, repeatable dispensing of reagents and other liquids. They serve as a very good alternative to pipettes or other conventional dispensing devices. They are suitable for most biotech, diagnostic and pharmaceutical applications.
Documents
WHEA211624_TDS_EN
Datasheet
Specifications
- Thread:8-425
- Capacity:3 ml
- Material:LDPE, translucent
- Seq:01
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



