Order Entry
Germany
Orders LinkContactUsLinkComponent
Bottles, narrow neck, with thread

Bottles, narrow neck, with thread

125 ml
Catalog # 215-2617
Supplier:   GRAVIS
Bottles, narrow neck, with thread
Catalog # 215-2617
Supplier:  GRAVIS
Bottles, narrow neck, with thread
Bottles, narrow neck, with thread

125 ml

Price
119,00 €
Log in for availability
1 * 104 ST
Product Details & Documents

Glass, clear, amber or yellow.

  • Neck Ø: 28 mm
  • Good chemical and temperature resistance
  • Amber coloured to protect light sensitive substances

Choice of various screw caps.

Ordering information: Supplied without screw caps - please order separately.

Packaging: Packaged in impermeable plastic packs.

Specifications
  • Capacity:
    125 ml
  • Seq:
    14
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.