Bottles, narrow neck, with thread
100 ml
215-2616
: GRAVIS
Glass, clear, amber or yellow.
- Neck Ø: 28 mm
- Good chemical and temperature resistance
- Amber coloured to protect light sensitive substances
Choice of various screw caps.
: Supplied without screw caps - please order separately.
: Packaged in impermeable plastic packs.
- Capacity:100 ml
- Seq:13



