ContactUsLinkComponent
The Plant Cell, 12 selected Microscope Slides of plant cytology - 1. Epidermis of Allium (onion), w.m. showing simple plant cells wi th cell walls, nuclei and cytoplasm 2. Root tips of Allium cepa l.s. showing cell division (mitosis) in all stages 3. 1 * 1 items
Catalog # LIED5100
Supplier:  LIEDER JOHANNES
The Plant Cell, 12 selected Microscope Slides of plant cytology - 1. Epidermis of Allium (onion), w.m. showing simple plant cells wi th cell walls, nuclei and cytoplasm 2. Root tips of Allium cepa l.s. showing cell division (mitosis) in all stages 3. 1 * 1 items
Catalog # LIED5100
Supplier:  LIEDER JOHANNES
Frequently Bought Together