Order Entry
Germany
Orders LinkContactUsLinkComponent
 

Electrolyte solutions, aqueous

Electrolyte solution, L 200 low temperature electrolyte (–30 °C), 1000 ml in DURAN® glass bottle, 1 L
Catalog # SCOI285138151
Supplier:   SI Analytics
Electrolyte solutions, aqueous
Catalog # SCOI285138151
Supplier:  SI Analytics
CAS Number:  
Electrolyte solutions, aqueous
Electrolyte solutions, aqueous

Electrolyte solution, L 200 low temperature electrolyte (–30 °C), 1000 ml in DURAN® glass bottle, 1 L

CAS Number:  
Price
149,00 €
Log in for availability
1 * 1 L
Product Details & Documents

Electrolyte aqueous solutions are used for reference electrodes, as electrolyte bridges and for storage.

Specifications

Specifications

  • Pk:
    1 L
  • Description:
    Electrolyte solution, L 200 low temperature electrolyte (–30 °C), 1000 ml in DURAN® glass bottle

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.