Methanol
5 L
Catalog # LBCL4561201
Marketsource
Supplier: LABC LABORTECHNIK
Methanol
Catalog # LBCL4561201
Supplier: LABC LABORTECHNIK
Marketsource
CAS Number: 67-56-1
Jump to:
Product Details & Documents
Synonyms: Wood alcohol, Methyl alcohol, Hydroxy methane, Carbinol
- CAS Number:67-56-1
- Formula:CH₃OH
- MDL Number:MFCD00004595
- Molecular Weight:32.04 g/mol
- Merck Index:13,05984
- Boiling Pt:64.6 °C (1013 hPa)
- Melting Pt:–98 °C
- Density:0.7918 g/cm³ (20 °C)
- Flash Pt:11 °C (closed cup)
- Storage Temperature:Ambient
- EINECS:200-659-6
- UN:1230
- ADR:3,II
- REACH:01-2119433307-44
Specifications
Specifications
- Pk:5 L
- Cat. No.:LBCL4561201
- Environmental certification:Chem21 Solvent Selection Guide recommended after discussion
- Environmentally Preferable:Safer Human and Environmental Health



