Order Entry
Germany
Orders LinkContactUsLinkComponent
 

Pycnometer heads with/without bottle

500 ml, 45/27
  LENZ05660006
Pycnometer heads with/without bottle
  LENZ05660006
Pycnometer heads with/without bottle
Pycnometer heads with/without bottle

500 ml, 45/27

 
89,10 €
   
1 * 1 ST
 

Set, made of DURAN® borosilicate glass, consists of wide neck bottle with conical shoulder and NS joint, and pycnometer head.

  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes

Suitable for density determinations of bitumen-like and mineral materials.

 
  • NS:
    45/27
  • Volume (nom.):
    500 ml
  • Seq:
    01
Accessories

 

  •  
    Description
     
     
  • LENZ07690070
    Bottle for pycnometer NS60/46, 1000 ml
       
    1 * 1 ST - 62,40 €