Orders LinkContactUsLinkComponent
Apparatus - Antonacopoulos
Spare distilling link with 2 cones, 29/32 NS, - NS
Catalog # LENZ05434004
Supplier:   Lenz Laborglas
Apparatus - Antonacopoulos
Catalog # LENZ05434004
Supplier:  Lenz Laborglas
Apparatus - Antonacopoulos
Spare distilling link with 2 cones, 29/32 NS, - NS
Price
62,40 €
Log in for availability
1 * 1 ST
Product Details & Documents
DURAN®, Borosilicate glass.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Specifications
  • Cone size:
    29/32 NS
  • Socket size:
    - NS
  • Description:
    Spare distilling link with 2 cones
  • Seq:
    04