Orders LinkContactUsLinkComponent
Corning® Microplate Lids
Black universal lid with corner notch, Sterile
Catalog # 734-4165
   Low Manufacturing Impact
Supplier:   Corning
Corning® Microplate Lids
Catalog # 734-4165
Supplier:  Corning
   Low Manufacturing Impact
Corning® Microplate Lids
Corning® Microplate Lids
Black universal lid with corner notch, Sterile
Price
130,00 €
Log in for availability
1 * 50 ST
Product Details & Documents
All lids are made of rigid polystyrene except where indicated.
All lids have a corner notch on the A1 corner (except where indicated) to correspond to the corner notches found on all Corning® microplates.
The universal lid without a corner notch (Cat. No. 3098) does not need to be oriented in any particular direction to be placed on Corning microplates. The lid also has a shorter skirt than standard lids.
The black universal lid (Cat. No. 3935) is suitable for fluorescent and other light-sensitive assays.
The DMSO-resistant cyclic olefin copolymer (COC) lid (Cat. No. 3085) is tinted amber in color for light-sensitive assays and is 100% DMSO-resistant.
Specifications
  • Sterility:
    Sterile
  • Description:
    Black universal lid with corner notch
  • Seq:
    04
  • Cat. No.:
    734-4165
  • Environmental certification:
    RECs, ISO 14001 - Kennebunk
  • Environmentally Preferable:
    Low Manufacturing Impact