Order Entry
Germany
Orders LinkContactUsLinkComponent
Saccharometers

Saccharometers

15 - 20 %, 0,1, 5…25 °C
Catalog # 641-0004
Supplier:   GERING
Saccharometers
Catalog # 641-0004
Supplier:  GERING
Saccharometers
Saccharometers

15 - 20 %, 0,1, 5…25 °C

Price
172,00 €
Log in for availability
1 * 1 ST
Product Details & Documents

These saccharometers with temperature correction scale are suitable for the official testing in breweries especially on wort.

Specifications
  • Temperature Range:
    5…25 °C
  • Seq:
    06
  • Division:
    0,1
  • Range:
    15 - 20 %