Order Entry
Germany
Orders LinkContactUsLinkComponent
Quinoline ≥98%

Quinoline ≥98%

Glass bottle for solids, 50 g
Catalog # 43225.18
Toxic
Health
Irritant
Quinoline ≥98%
Catalog # 43225.18
CAS Number:  91-22-5
Quinoline ≥98%
Quinoline ≥98%

Glass bottle for solids, 50 g

CAS Number:  91-22-5
Price
34,70 €
Log in for availability
1 * 50 g
Product Details & Documents

Purity: 98% CAS No.: 91-22-5 Molecular Formula: C9H7N Formula Weight: 129.16 Flash Point: 101°(214°F) Physical Form: Liquid Density: 1.090 Refractive Index: 1.6260 Sensitivity: Light Sensitive and Hygroscopic Appearance: Colourless to pale yellow pH: 7.3 Melting Point: -15° Boiling Point: 237° Beilstein Reference: 107477 Merck Reference: 14,8068 UN No.: 2656 MDL No.: MFCD00006736

  • CAS Number:91-22-5
  • Formula:C₉H₇N
  • MDL Number:MFCD00006736
  • Molecular Weight:129.16 g/mol
  • Merck Index:14,8068
  • Boiling Pt:237 °C
  • Melting Pt:–15 °C
  • Density:1.090
  • Flash Pt:101 °C (214 °F)
  • Storage Temperature:Ambient
  • EINECS:202-051-6
  • UN:2656
  • ADR:6.1,III
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    50 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.