Multielement standard solution 17 elements in nitric acid approx. 1 mol/l 1 Liter contains: 500 mg: Ca, K, Mg, Na 50 mg: Al, Cd, Co, Cr, Cu, Fe, Mn, Ni, Pb,... 1 * 500 mL
KRAF02426.2000
Multielement standard solution 17 elements in nitric acid approx. 1 mol/l 1 Liter contains: 500 mg: Ca, K, Mg, Na 50 mg: Al, Cd, Co, Cr, Cu, Fe, Mn, Ni, Pb,... 1 * 500 mL
KRAF02426.2000
Frequently Bought Together
Acetone ≥99.5%, GPR RECTAPUR®
: VWR Chemicals
Acetone ≥99%, TECHNICAL
: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
Ethanol 96% ≥96% (v/v), TechniSolv®, pure
: VWR Chemicals
Diethyl ether ≥99% stabilised, GPR RECTAPUR®
: VWR Chemicals