POTASSIUM IODOBISMUTHATE SOLUTION 1 * 100 mL
Catalog # REGE1070600
Lieferant: REAGECON
Frequently Bought Together
Detergents, liquid, for automated cleaning of glassware, neodisher® FLA
Lieferant: Dr. Weigert
Acetone ≥99.5%, GPR RECTAPUR®
Lieferant: VWR Chemicals
Acetone ≥99%, TECHNICAL
Lieferant: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Lieferant: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
Lieferant: VWR Chemicals
Neutralising agent for the automated cleaning of glassware, neodisher® Z
Lieferant: Dr. Weigert
Syringe without needle TERUMO 5.0ml- Slip Tip-Centric 1 * 100 items
Lieferant: TERUMO
VWR® Facial Tissues
Lieferant: VWR Collection
VWR®, Nitrile Gloves
Lieferant: VWR Collection
VWR® Mercuric Bromide Paper (Reag. Ph. Eur 9. 1052101) for the Determination of Arsenic
Lieferant: VWR Chemicals
VWR®, Transfer Pipettes, High Performance
Lieferant: VWR Collection
VWR® NITRILE LIGHT, Nitrile Gloves
Lieferant: VWR Collection
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Lieferant: VWR Chemicals
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Lieferant: VWR Chemicals