Order Entry
Germany
Orders LinkContactUsLinkComponent
Test fluids for viscometers

Test fluids for viscometers

100BW DKD Certificated, 100, 1
Catalog # HAAK082-5303
Supplier:   HAAKE
Test fluids for viscometers
Catalog # HAAK082-5303
Supplier:  HAAKE
CAS Number:  
Test fluids for viscometers

100BW DKD Certificated, 100, 1

CAS Number:  
Price
395,00 €
Log in for availability
1 * 1 ST
Product Details & Documents

Test and certificated fluids for viscometery - mesurement temperature at 20 °C unless indicated.

Caution: * Measurement temperature 25 °C

Documents

Specifications

Specifications

  • Pk:
    1
  • Type:
    100BW DKD Certificated
  • Viscosity:
    100

Specifications