Microscope slide box, 25place, cork lined, blue 1 * 72 items
Catalog # 631-9089
Discontinued
Product has been discontinued. However we found similar alternative products.
Alternatives
SLIDE BOX 25-PL BLUE CORK LINED 1 * 1 items Catalog #631-1506
Product 631-1506 is similar to 631-9089 but may have a different manufacturer and/or packaging.
slide Box for 25 slides, white, with snap lock, made of polystyrene, use in freezer down to -80°c and usable up to +70°c, for slides 76 mm x 26 mm x 1 mm 1 * 1 items Catalog #GLWAK25B
Product GLWAK25B is similar to 631-9089 but may have a different manufacturer and/or packaging.