Orders LinkContactUsLinkComponent
Measuring cup, disposable
Measuring cup, disposable
Measuring cup EMB-Z2
Catalog # PAAR301510
Supplier:   Anton Paar
Measuring cup, disposable
Catalog # PAAR301510
Supplier:  Anton Paar
Measuring cup, disposable
Measuring cup EMB-Z2
Price
918,00 €
Log in for availability
1 * 100 ST
Product Details & Documents
Cup made of aluminum.
  • Good chemical resistance
  • Good temperature resistance
  • Disposable
Specifications
  • Description:
    Measuring cup EMB-Z2
  • Seq:
    01