ContactUsLinkComponent
Buffer solution pH 1054 g NH4Cl/l + 350 ml NH3-solution 25 %/l for complexometric titration Content: 1 l 1 * 1 L
Catalog # KRAF03111.3000
Supplier:  Bernd Kraft
Buffer solution pH 1054 g NH4Cl/l + 350 ml NH3-solution 25 %/l for complexometric titration Content: 1 l 1 * 1 L
Catalog # KRAF03111.3000
Supplier:  Bernd Kraft