ContactUsLinkComponent
o-Tolidine solution 1.35 g o-Tolidine dihydrochloride + 125 ml HCl 37 %/lin water for photometric determination of active chlorine C ontent: 0,5 l 1 * 500 mL
Catalog # KRAF03061.2000
Supplier:  Bernd Kraft
o-Tolidine solution 1.35 g o-Tolidine dihydrochloride + 125 ml HCl 37 %/lin water for photometric determination of active chlorine C ontent: 0,5 l 1 * 500 mL
Catalog # KRAF03061.2000
Supplier:  Bernd Kraft